Learn More
MSC Wearwell NBR Male Edging, 39 in, Yellow, 1 ea
Catalog No. 5000782628
Encompass
This male edging securely interlocks with 24/Seven NBR rubber floor tiles, facilitating a smooth transition for foot traffic. Made from Nitrile Rubber (NBR), it is specifically designed for optimal performance in environments with petroleum-based fluids and is suitable for both wet and dry conditions.
  • Securely interlocks with modular tiles
  • Yellow color provides visual cue for safety
  • Designed for areas with petroleum-based fluids
  • Chemical and abrasion-resistant
  • Resistant to cutting fluids
  • Suitable for wet and dry environments
Catalog No. 50-007-82628 Supplier MSC Supplier No. 84809235
This male edging securely interlocks with 24/Seven NBR rubber floor tiles, facilitating a smooth transition for foot traffic. Made from Nitrile Rubber (NBR), it is specifically designed for optimal performance in environments with petroleum-based fluids and is suitable for both wet and dry conditions.
  • Securely interlocks with modular tiles
  • Yellow color provides visual cue for safety
  • Designed for areas with petroleum-based fluids
  • Chemical and abrasion-resistant
  • Resistant to cutting fluids
  • Suitable for wet and dry environments