Learn More
MSC Wearwell Ramp Edge Mat, Anti-Fatigue, Dry and Wet, 1 ea
Catalog No. 5000793915
Encompass
This anti-fatigue modular ramp edge mat is engineered for demanding industrial environments, providing a safe transition from floor to mat in both wet and dry conditions. Its durable rubber construction offers robust resistance to abrasion, chemicals, and cutting fluids, ensuring long-lasting performance.
  • Designed for heavy-duty traffic
  • Features female connectors for modular matting tiles
  • Compatible with Series 570, 572, 574, and 576 modular matting tiles
  • Resistant to abrasion, chemicals, and cutting fluids
  • Provides a smooth transition for foot traffic and acts as a visual cue
  • Constructed from grease-resistant (GR) rubber, suitable for areas with water or dry debris
  • Made from recycled content
Catalog No. 50-007-93915 Supplier MSC Supplier No. 62686241
This anti-fatigue modular ramp edge mat is engineered for demanding industrial environments, providing a safe transition from floor to mat in both wet and dry conditions. Its durable rubber construction offers robust resistance to abrasion, chemicals, and cutting fluids, ensuring long-lasting performance.
  • Designed for heavy-duty traffic
  • Features female connectors for modular matting tiles
  • Compatible with Series 570, 572, 574, and 576 modular matting tiles
  • Resistant to abrasion, chemicals, and cutting fluids
  • Provides a smooth transition for foot traffic and acts as a visual cue
  • Constructed from grease-resistant (GR) rubber, suitable for areas with water or dry debris
  • Made from recycled content