Learn More
MSC Wearwell Rejuvenator Squared Runner End Female, 1 ea.
Catalog No. 5000685539
Encompass
The Wearwell Rejuvenator Squared Runner End Female mat is designed for creating custom-length ergonomic matting runs with a clean aesthetic. Featuring molded-in ramps, this mat reduces installation time by eliminating the need for separate edging. Its positive interlocking system beneath the surface allows for flexible configuration and replacement in dry environments.
  • Meets ASTM G21 standard for anti-microbial properties
  • Constructed from 100% urethane with a thick outer wear layer
  • Overall thickness is 5/8 in. (16 mm)
  • Interlocking modular design for on-site customization
  • Exclusive urethane connector system
Catalog No. 50-006-85539 Supplier MSC Supplier No. 74166349
The Wearwell Rejuvenator Squared Runner End Female mat is designed for creating custom-length ergonomic matting runs with a clean aesthetic. Featuring molded-in ramps, this mat reduces installation time by eliminating the need for separate edging. Its positive interlocking system beneath the surface allows for flexible configuration and replacement in dry environments.
  • Meets ASTM G21 standard for anti-microbial properties
  • Constructed from 100% urethane with a thick outer wear layer
  • Overall thickness is 5/8 in. (16 mm)
  • Interlocking modular design for on-site customization
  • Exclusive urethane connector system