Learn More
Wearwell™ Diamond-Plate Modular Mat Systems
Catalog No. 19034144 Shop All Wearwell Tennessee Mat Company Inc. Products
Change view
Click to view available options
Resistance:
Grease-resistant
Greaseproof
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Resistance
19-034-144 Grease-resistant
19-034-142 Greaseproof
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-034-144 Supplier Wearwell™ Supplier No. 470.12X3X3GRBK
Interlocking system ideal for use in single work spaces or large areas
  • Designed for comfort and ease of use in mostly dry areas
  • Interlocking system requires no additional bevels or attachments
  • Lightweight, easy to clean
  • Rounded exterior edges
  • Natural rubber for use in general applications
  • Grease-proof, 50% nitrile rubber for use in areas with occasional exposure to chemicals or caustic agents
Order Info
Thickness: 0.5 in. (1.3cm), dimensions: L x W: 3 x 3 ft. (91.4 x 91.4cm).
Specifications
Dimensions (L x W) 3 x 3 ft.
Resistance Grease-resistant