Learn More
MSC Wearwell UltraSoft Diamond-Plate, 15/16 in, 3x20 ft, 1 ea
Catalog No. 5000795211
Encompass
Wearwell UltraSoft Diamond-Plate SpongeCote anti-fatigue mats are engineered for high-performance industrial environments. These mats provide exceptional comfort and support, reducing fatigue in standing work applications through their advanced Nitricell sponge technology.
  • Outstanding fatigue relief
  • Longer lifespan than alternatives
  • Customizable shapes and sizes
  • Diamond-plate PVC tread for improved traction
  • Made from 100% post-industrial recycled material
  • Nitricell sponge is 2-3x more resilient than PVC sponge
  • Closed-cell Nitricell minimizes fluid absorption
  • Uniform thickness with precision skiving
  • Stimulates muscle activity, improves circulation
  • UltraSoft 15/16 in thick Nitricell for maximum comfort
  • NFSI certified
  • Pre-beveled edges prevent tripping
Catalog No. 50-007-95211 Supplier MSC Supplier No. 91834671
Wearwell UltraSoft Diamond-Plate SpongeCote anti-fatigue mats are engineered for high-performance industrial environments. These mats provide exceptional comfort and support, reducing fatigue in standing work applications through their advanced Nitricell sponge technology.
  • Outstanding fatigue relief
  • Longer lifespan than alternatives
  • Customizable shapes and sizes
  • Diamond-plate PVC tread for improved traction
  • Made from 100% post-industrial recycled material
  • Nitricell sponge is 2-3x more resilient than PVC sponge
  • Closed-cell Nitricell minimizes fluid absorption
  • Uniform thickness with precision skiving
  • Stimulates muscle activity, improves circulation
  • UltraSoft 15/16 in thick Nitricell for maximum comfort
  • NFSI certified
  • Pre-beveled edges prevent tripping