Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Weather Stencils
SDPSCI_ED

Catalog No. S13226
Change view
Click to view available options
Grade:
1+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S13226 1+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13226 Supplier Roylco™ Supplier No. R58630
Add to Cart
Add to Cart

Introduce and explore weather events through art with these weather stencils!

  • Quality stencils are durable and reusable for years
  • Create pictures of the seasons, or to help illustrate lessons about the weather and the environment
  • Simply punch out the designs and start creating
  • Trace the patterns on paper and color them in or cut out the tracings to build a collage
  • Trace them over t-shirts or sidewalks, dry erase boards and more
  • Use your stencils with paints, inks, pastels, pencils, crayons, or markers
  • Stencils are washable and reusable
  • Promotes STEAM and fine motor skills, the art and science intersection
  • Dimensions: 5 x 9 in. (13 x 23 cm)

Specifications

Grade Level Elementary, Intermediate - Middle School, High School
Grade 1+
Format Kit
Quantity 10 Stencils
Languages English, Spanish, French, German
Subject Earth Science - Weather and Climate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.