Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AquaPhoenix™ BCPS Deli Container
SCI_ED

Catalog No. S39380KWW Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
S39380KWW 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39380KWW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deli container replacement parts for educational kit.

  • Variety of components
  • Specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits

Specifications

Grade K to 5
For Use With (Application) Education Kit
Capacity (English) 16 oz.
Product Type Deli Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.