Learn More
Mfi Medical Weaver Nuprep Skin Prep Gel, 4oz tubes, 3/pk
SDP
Catalog No. NC1594379
Encompass
Weaver Nuprep Skin Prep Gel effectively lowers impedance to improve your tracings. Its mild abrasive formula improves conductivity and helps achieve maximum efficiency with your equipment.
Catalog No. NC1594379 Supplier Mfi Medical Supplier No. WVR1030
Weaver Nuprep Skin Prep Gel effectively lowers impedance to improve your tracings. Its mild abrasive formula improves conductivity and helps achieve maximum efficiency with your equipment.