Learn More
MSC Lift-All Endless Web Sling, Polyester, 4 in., 14 ft., 11,500 lb. Vertical Capacity, 1 ea.
Catalog No. 5000510908
Encompass
This Type 5 endless web sling is a heavy-duty polyester lifting solution designed for versatile material handling applications. The sling features a 4 in. width and a 14 ft. length, engineered to provide reliable performance in vertical, choker, and basket hitch configurations. With a vertical capacity of 11,500 lbs., this sling offers durability and load control for industrial lifting tasks.
  • Polyester webbing stretches less for better load control
  • Yellow abrasion-resistant treatment extends service life
  • Red core warning yarns aid in the inspection process
  • Includes Tuff-Tag for permanent identification as required by OSHA
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Operating temperature range of -60°F to 200°F
  • Reinforced with abrasion- and cut-resistant edges
Catalog No. 50-005-10908 Supplier MSC Supplier No. 87843280
This Type 5 endless web sling is a heavy-duty polyester lifting solution designed for versatile material handling applications. The sling features a 4 in. width and a 14 ft. length, engineered to provide reliable performance in vertical, choker, and basket hitch configurations. With a vertical capacity of 11,500 lbs., this sling offers durability and load control for industrial lifting tasks.
  • Polyester webbing stretches less for better load control
  • Yellow abrasion-resistant treatment extends service life
  • Red core warning yarns aid in the inspection process
  • Includes Tuff-Tag for permanent identification as required by OSHA
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
  • Operating temperature range of -60°F to 200°F
  • Reinforced with abrasion- and cut-resistant edges