Learn More
MSC Lift-All Web Sling, Universal Link, Polyester, 1 Ply, 1 ea
Catalog No. 5000515592
Encompass
The universal link web sling is a 1-ply polyester lifting sling designed for general-purpose, moderate-duty applications. It offers reliable load control with minimal stretch and includes integrated resistance to grease and most acids. For enhanced safety, the sling features a red core warning yarn system that visually indicates damage or excessive wear, signaling the need for immediate replacement.
  • Rated vertical capacity of 4,800 lb., choker capacity of 3,800 lb., and basket capacity of 9,600 lb.
  • Abrasion-resistant polyester construction with latex yarn coating.
  • Operating temperature range from -60°F to 200°F.
  • Includes TUFF-TAG™ for durability and identification.
  • Compliant with OSHA 1910.184, ASME B30.9, and WSTDA standards.
  • Single-ply, 3 in. wide webbing with a 3 ft. length.
Catalog No. 50-005-15592 Supplier MSC Supplier No. 88360649
The universal link web sling is a 1-ply polyester lifting sling designed for general-purpose, moderate-duty applications. It offers reliable load control with minimal stretch and includes integrated resistance to grease and most acids. For enhanced safety, the sling features a red core warning yarn system that visually indicates damage or excessive wear, signaling the need for immediate replacement.
  • Rated vertical capacity of 4,800 lb., choker capacity of 3,800 lb., and basket capacity of 9,600 lb.
  • Abrasion-resistant polyester construction with latex yarn coating.
  • Operating temperature range from -60°F to 200°F.
  • Includes TUFF-TAG™ for durability and identification.
  • Compliant with OSHA 1910.184, ASME B30.9, and WSTDA standards.
  • Single-ply, 3 in. wide webbing with a 3 ft. length.