Learn More
MSC Lift-All Endless Web Sling, Polyester, 3 in. Wide, 10 ft. Long, 1 ea.
Catalog No. 5000515338
Encompass
This endless, Type 5 web sling is designed for general-purpose lifting, providing a 16,300 lb vertical load capacity. Constructed from durable, abrasion-resistant polyester, the sling is suitable for daily use under rugged lifting conditions. It features a versatile design that accommodates vertical, choker, and basket hitches, allowing the sling to be rotated to minimize localized wear.
  • Features 16,300 lb vertical, 13,000 lb choker, and 32,600 lb basket hitch capacities
  • Constructed from heavy-duty, 2-ply polyester with abrasion-resistant edges
  • Operating temperature range from -60°F to 200°F
  • Visual damage indicators with red core yarns to alert users to potential strength loss
  • Includes safety bulletin
  • Compliant with WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-15338 Supplier MSC Supplier No. 87844072
This endless, Type 5 web sling is designed for general-purpose lifting, providing a 16,300 lb vertical load capacity. Constructed from durable, abrasion-resistant polyester, the sling is suitable for daily use under rugged lifting conditions. It features a versatile design that accommodates vertical, choker, and basket hitches, allowing the sling to be rotated to minimize localized wear.
  • Features 16,300 lb vertical, 13,000 lb choker, and 32,600 lb basket hitch capacities
  • Constructed from heavy-duty, 2-ply polyester with abrasion-resistant edges
  • Operating temperature range from -60°F to 200°F
  • Visual damage indicators with red core yarns to alert users to potential strength loss
  • Includes safety bulletin
  • Compliant with WSTDA, ASME B30.9, and OSHA 1910.184 standards