Learn More
MSC Lift-All Universal Link Web Sling, 3 in. Wide, 4 ft. Long, Polyester, 1 ea
Catalog No. 5000515342
Encompass
This Universal Link, Type U web sling is a heavy-duty, one-ply polyester lifting tool designed for versatile performance in vertical, choker, and basket hitch configurations. It features durable Webmaster 1600 polyester webbing with an abrasion-resistant latex coating, providing reliable strength for industrial lifting applications.
  • Versatile universal link fittings on each end allow for efficient choking and varied wear points.
  • High-strength polyester construction supports a vertical load capacity of 4,800 lb.
  • Reinforced with cut-resistant and abrasion-resistant edges for enhanced durability.
  • Steel fittings provide secure attachment points in multiple hitch types.
  • Operates effectively in a wide temperature range from -60°F to 200°F.
  • Compliant with OSHA 1910.184, ASME B30.9, and WSTDA standards.
Catalog No. 50-005-15342 Supplier MSC Supplier No. 88360656
This Universal Link, Type U web sling is a heavy-duty, one-ply polyester lifting tool designed for versatile performance in vertical, choker, and basket hitch configurations. It features durable Webmaster 1600 polyester webbing with an abrasion-resistant latex coating, providing reliable strength for industrial lifting applications.
  • Versatile universal link fittings on each end allow for efficient choking and varied wear points.
  • High-strength polyester construction supports a vertical load capacity of 4,800 lb.
  • Reinforced with cut-resistant and abrasion-resistant edges for enhanced durability.
  • Steel fittings provide secure attachment points in multiple hitch types.
  • Operates effectively in a wide temperature range from -60°F to 200°F.
  • Compliant with OSHA 1910.184, ASME B30.9, and WSTDA standards.