Learn More
MSC Lift-All Endless Web Sling, Polyester, 4 in. x 10 ft., 20,700 lb. Vertical Capacity, 1 ea.
Catalog No. 5000510962
Encompass
This Type 5 endless web sling is designed for heavy-duty lifting applications, providing a vertical capacity of 20,700 lbs. Engineered with 2-ply polyester webbing, this sling features low stretch characteristics for enhanced load control. The durable construction is rated for temperatures ranging from -60°F to 200°F and meets WSTDA, ASME B30.9, and OSHA 1910.184 safety standards.
  • Yellow abrasion treatment extends the service life of the sling
  • Red core warning yarns aid in the inspection process
  • Includes Tuff-Tag for permanent identification as required by OSHA
  • Versatile design supports vertical, choker, and basket hitches
  • Abrasion and cut-resistant edges improve durability in rugged environments
Catalog No. 50-005-10962 Supplier MSC Supplier No. 88183421
This Type 5 endless web sling is designed for heavy-duty lifting applications, providing a vertical capacity of 20,700 lbs. Engineered with 2-ply polyester webbing, this sling features low stretch characteristics for enhanced load control. The durable construction is rated for temperatures ranging from -60°F to 200°F and meets WSTDA, ASME B30.9, and OSHA 1910.184 safety standards.
  • Yellow abrasion treatment extends the service life of the sling
  • Red core warning yarns aid in the inspection process
  • Includes Tuff-Tag for permanent identification as required by OSHA
  • Versatile design supports vertical, choker, and basket hitches
  • Abrasion and cut-resistant edges improve durability in rugged environments