Learn More
Enzo Life Sciences WEB-2086 (5mg). CAS: 105219-56-5
SDP
Supplier:  Enzo Life Sciences BMLL1370005
Encompass_Preferred
PAF receptor antagonist. Inhibits PAF-induced human platelet and neutrophil aggregation in vitro (IC50=0.17 and 0.36µM respectively). In vivo it dose dependently blocks eosinophil activation and displays antiallergic and anti-inflammatory activity in various models. Induces differentiation and triggers apoptosis and in a variety of cell lines. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO or ethanol. Long Term Storage: +4°C.
Catalog No. 50-201-1139