Learn More
MSC Lift-All Web Sling, Webmaster 1200, Polyester, 1-Ply, Flat Eyes
Catalog No. 5000513512
Encompass
The Lift-All Webmaster 1200 Web Sling is a durable and reliable tool designed for lifting applications. Constructed from 1-ply polyester, it features a flat eyes design for secure handling and easy rigging. This sling is equipped with a red core yarn warning system for enhanced safety and is resistant to rot and mildew, making it suitable for various industrial environments.
  • Tuff-Tag rated capacity and warning label guaranteed legible
  • Red core yarn warning system aids in sling inspection
  • Abrasion resistant latex treatment for extended sling life
  • Soft web will not mar the load
  • Resists rot and mildew
  • Flat eyes (type 3) for easy rigging and removal from loads
  • Chokes tightly around loads
Catalog No. 50-005-13512 Supplier MSC Supplier No. 01136902
The Lift-All Webmaster 1200 Web Sling is a durable and reliable tool designed for lifting applications. Constructed from 1-ply polyester, it features a flat eyes design for secure handling and easy rigging. This sling is equipped with a red core yarn warning system for enhanced safety and is resistant to rot and mildew, making it suitable for various industrial environments.
  • Tuff-Tag rated capacity and warning label guaranteed legible
  • Red core yarn warning system aids in sling inspection
  • Abrasion resistant latex treatment for extended sling life
  • Soft web will not mar the load
  • Resists rot and mildew
  • Flat eyes (type 3) for easy rigging and removal from loads
  • Chokes tightly around loads