Learn More
MSC Lift-All Web Sling, Polyester, Twisted Eye, 2 Ply, 14 ft., 1 ea.
Catalog No. 5000515653
Encompass
The Webmaster 1600 twisted eye and eye web sling is a heavy-duty polyester sling designed for reliable lifting applications. Featuring a 2-ply construction, this sling provides exceptional strength and durability for various rigging needs. It is built to operate safely in temperature ranges from -60°F to 200°F and is engineered to resist both abrasion and cuts.
  • Features a soft twill weave body for flexible handling.
  • Includes a red core yarn warning system to aid in the inspection process.
  • Yellow treatment provides superior abrasion resistance to extend sling life.
  • Identified by a single blue surface stripe.
  • Constructed with a tubular design and reinforced core.
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA standards.
Catalog No. 50-005-15653 Supplier MSC Supplier No. 81194060
The Webmaster 1600 twisted eye and eye web sling is a heavy-duty polyester sling designed for reliable lifting applications. Featuring a 2-ply construction, this sling provides exceptional strength and durability for various rigging needs. It is built to operate safely in temperature ranges from -60°F to 200°F and is engineered to resist both abrasion and cuts.
  • Features a soft twill weave body for flexible handling.
  • Includes a red core yarn warning system to aid in the inspection process.
  • Yellow treatment provides superior abrasion resistance to extend sling life.
  • Identified by a single blue surface stripe.
  • Constructed with a tubular design and reinforced core.
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA standards.