Learn More
MSC Lift-All Web Sling, Type 3, 2 Ply, Polyester, 16 ft. x 1 in., 1 ea.
Catalog No. 5000515411
Encompass
This Type 3 flat eye and eye web sling is engineered for versatile material handling and lifting applications. Constructed from high-strength polyester, this 2-ply sling offers reliable performance and durability in industrial environments. It is designed to meet rigorous safety standards and includes integrated features to assist with inspection and longevity.
  • Features 2-ply polyester construction with a 16 ft. length and 1 in. width
  • Provides a vertical capacity of 3,200 lbs., choker capacity of 2,500 lbs., and basket capacity of 6,400 lbs.
  • Includes blue polymer edge yarns for enhanced cut resistance
  • Features a silver color treatment for improved abrasion resistance
  • Equipped with red core warning yarns to indicate web damage
  • Includes a permanent identification tag as required by OSHA
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards
Catalog No. 50-005-15411 Supplier MSC Supplier No. 85813590
This Type 3 flat eye and eye web sling is engineered for versatile material handling and lifting applications. Constructed from high-strength polyester, this 2-ply sling offers reliable performance and durability in industrial environments. It is designed to meet rigorous safety standards and includes integrated features to assist with inspection and longevity.
  • Features 2-ply polyester construction with a 16 ft. length and 1 in. width
  • Provides a vertical capacity of 3,200 lbs., choker capacity of 2,500 lbs., and basket capacity of 6,400 lbs.
  • Includes blue polymer edge yarns for enhanced cut resistance
  • Features a silver color treatment for improved abrasion resistance
  • Equipped with red core warning yarns to indicate web damage
  • Includes a permanent identification tag as required by OSHA
  • Complies with WSTDA, ASME B30.9, and OSHA 1910.184 standards