Learn More
MSC Lift-All Webmaster 1600 Web Sling, Polyester, 3 in, 14 ft, 4,800 lb, 1 ea
Catalog No. 5000512615
Encompass
This heavy-duty Type 3 flat eye and eye web sling is designed for versatile use in vertical, choker, and basket hitches. Constructed from durable polyester, it offers high strength, minimal stretch for load control, and resistance to UV light, grease, and most acids. Its flexible design and wide bearing surface help protect the load during lifting operations.
  • Features a red core yarn warning system to aid in inspection
  • Treated with a yellow coating for enhanced abrasion resistance and extended service life
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA standards
  • Includes reinforced edges for cut and abrasion protection
  • Suitable for general purpose lifting applications
Catalog No. 50-005-12615 Supplier MSC Supplier No. 76691351
This heavy-duty Type 3 flat eye and eye web sling is designed for versatile use in vertical, choker, and basket hitches. Constructed from durable polyester, it offers high strength, minimal stretch for load control, and resistance to UV light, grease, and most acids. Its flexible design and wide bearing surface help protect the load during lifting operations.
  • Features a red core yarn warning system to aid in inspection
  • Treated with a yellow coating for enhanced abrasion resistance and extended service life
  • Complies with OSHA 1910.184, ASME B30.9, and WSTDA standards
  • Includes reinforced edges for cut and abrasion protection
  • Suitable for general purpose lifting applications