Learn More
Thermo Scientific™ WebREMS™ Web-based Real-time Environmental Monitoring System
Catalog No. 19175202
Change view
Click to view available options
Product Type:
Single Site License
Site License
For Use With (Application):
0 to 100 Devices
0 to 1500 Devices
0 to 200 Devices
0 to 50 Devices
0 to 750 Devices
Unlimited
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
19-175-202 Single Site License 0 to 50 Devices
19-175-203 Single Site License 0 to 100 Devices
19-175-204 Single Site License 0 to 200 Devices
19-175-205 Single Site License 0 to 750 Devices
19-175-206 Single Site License 0 to 1500 Devices
19-175-209 Site License Unlimited
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-175-202 Supplier Thermo Scientific™ Supplier No. SYS/1/70860/002
Only null left
Add to Cart
Add to Cart
Request a Quote
Be sure personnel exposed to ionizing radiation don't exceed allowable dose limits and receive appropriate decontamination treatment with a web-based EPD monitoring system.
Personnel working in environments where they are exposed to ionizing radiation must have their exposure accurately monitored, recorded and reported. The Thermo Scientific™ webREMS™ Web-based Real-time Environmental Monitoring System tracks vital data from your electronic personal dosimeter (EPD) population including worker dosimeter visits, training status, dosimeters status, tasks\permits and radiation dose summary. The system also performs the EPD issue and return process. With this easy-to-use web-based graphical interface, accurate data and full reporting capabilities are at your fingertips.
Thermo Scientific webREMS Web-based Real-Time Environmental Monitoring System for monitoring and reporting personnel radiation exposure has built-in reporting capability and allows system administration from anywhere in the world.
Features
  • Client/server web-based application designed to track and monitor personnel radiation exposure.
  • Facilitates distributed access control through simple to manage electronic personal dosimeter (EPD) issue and return stations through an easy to use web browser, utilizing Microsoft™ Internet Explorer™ (IE).
  • Facilitates data management of personnel radiological dose limits, training requirements and authorization against Tasks.
  • Integrates with Thermo Scientific™ ViewPoint™ Enterprise to provide real-time monitoring of personnel exposed to ionizing radiation.
  • Integrates with Thermo Scientific™ webALARA web-based As Low As Reasonably Achievable permit management system.
  • Provides reporting capabilities of all visits performed using the system, recording and reporting the dose recorded against personnel and tasks/permits over one or more configurable time periods.
  • Facilitates simple, quick integration with existing data through a built-in import process for personnel, dosimeter calibration expiry and training data.
  • Scalable such that webREMS may be installed and operated from a single laptop PC up to an enterprise server installation providing access anywhere in the world.
  • Secure (HTTPS) access over public internet with password protected system access via ViewPoint Authentication Service.
  • Typical uses include nuclear power plants, government labs, federal agencies, security and military.
Requires
Microsoft™ Windows™ PC with Internet Explorer™ (IE) and Internet Information Services (IIS).
Thermo Scientific™ Electronic Personal Dosimeter (EPD), Beta/Gamma, Gamma, Neutron or Fast-Neutron.
Thermo Scientific™ IRDA Reader to use Access Control.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Single Site License
Compatibility Thermo Scientific EPD Mk2, EPD-N, EPD-N2
Operating System Microsoft Windows 2008, 2008 R2, XP, 7 (32 and 64 bit)
For Use With (Application) 0 to 50 Devices