Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WebSeal™ CLR Silicone Mat, 384 square wells
GSA_VA

Catalog No. p-8015794 Shop All Thermo Scientific Products
Change view
Click to view available options
Pre-slit:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit
03-252-438 No
03-252-439 Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-252-438 Supplier Thermo Scientific™ Supplier No. 60180M126
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eliminate cross contamination and ensure an inert and safe seal of your plates, every time. Mat materials resist coring and tearing and also offer superior resealability.

Store at room temperature

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material Silicone
Sterility Non-sterile
No. per Pack 5
Pre-slit No
Well Shape Square
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.