Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Premier Webster Needle Holder

Catalog No. 22079757 Shop All Surgical Design Products
Change view
Click to view available options
Length (English):
5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22-079-757 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-079-757 Supplier Surgical Design™ Supplier No. GM20304S
Add to Cart
Add to Cart

Surgical Design™ Premier Grade Webster Needle Holder is used for driving small needles and fine suture material.

  • Ultra-smooth, beveled-edge finish
  • Snag-Free
  • Guaranteed free from defect
  • Large rings for comfort and ease of use
  • Corrosion protected by passivation and ultrasonic cleaning
  • Properly tempered - Rockwell Hardness (HRC): 40-45
  • Sterilizable by EtOH/Gamma Radiation
  • Fully autoclavable

Specifications

Product Type Needle Driver
Autoclavable Autoclavable
Color Silver
Material Premium Stainless Steel
For Use With (Application) Needle Driving
Grade Surgical
Latex Free Yes
Quantity 1
Length (English) 5 in.
Length (Metric) 12.7 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.