Learn More
World Precision Instrument Webster Needle Holder, 14cm, Straight, Serrated, Extra Delicate, Ring Lock Mechanism
SDP
Catalog No. 502521374
Encompass
Webster needle holders are surgical instrument use to hold and manipulate suturing needles. While Webster needle holders are often associated with microsurgery and microvascular procedures, they can be used in a variety of surgical contexts where precise needle handling is required. Webster needle holders have fine tips for precise control when grasping needles. The fine tips make it easier to handle and manipulate small and fine needles used in microsurgery and microvascular procedures. We also offer extra fine tips for smaller needles. Like most needle holders, Webster needle holders have a locking handle for a secure hold on the needle, so that it remains stable during suturing, reducing the risk of slipping or accidental needle sticks. The jaws of some WPI Webster needle holders are lightly textured for a solid grip. Others have smooth jaws to minimize tissue damage. .
Catalog No. 50-252-1374 Supplier World Precision Instrument Supplier No. 501929
Webster needle holders are surgical instrument use to hold and manipulate suturing needles. While Webster needle holders are often associated with microsurgery and microvascular procedures, they can be used in a variety of surgical contexts where precise needle handling is required. Webster needle holders have fine tips for precise control when grasping needles. The fine tips make it easier to handle and manipulate small and fine needles used in microsurgery and microvascular procedures. We also offer extra fine tips for smaller needles. Like most needle holders, Webster needle holders have a locking handle for a secure hold on the needle, so that it remains stable during suturing, reducing the risk of slipping or accidental needle sticks. The jaws of some WPI Webster needle holders are lightly textured for a solid grip. Others have smooth jaws to minimize tissue damage. .