Learn More
Sklar Instruments Surgi-OR Webster Needle Holder, Sklar 5-1/4 in., Straight, Serrated, Stainless Steel. EA.
SDP
Catalog No. 501184243 Shop All Sklar Instruments Products
Encompass_Preferred
Surgi-OR Webster Needle Holder, Sklar. 5-1/4 in., Straight, Serrated, Stainless Steel. Surgi-OR TC Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with serrated tips and a length of 5-1/4 inches.
Catalog No. 50-118-4243 Supplier Sklar Instruments Supplier No. 95813
Surgi-OR Webster Needle Holder, Sklar. 5-1/4 in., Straight, Serrated, Stainless Steel. Surgi-OR TC Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures. They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Hand-crafted in Pakistan, Surgi-OR TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with serrated tips and a length of 5-1/4 inches.