Learn More
Electron Microscopy Sciences K. WecPrep Blades, single edge, 50/pk
SDP
Supplier:  Electron Microscopy Sciences 7193350
Encompass_Preferred
Single edge, carbon steel razor blades which are long and very stable. Used for mincing and all other specimen preparation work. Blade Width: 2.25" (57.15 mm); Blade Thickness: .011" (.279 mm)
Catalog No. 50-949-469