Learn More
TARGETMOL CHEMICALS INC WEDELOLACTONE 10MG
SDP
Catalog No. 502576953
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Wedelolactone (IKK Inhibitor II) inhibits NF-(kappa)B-mediated gene transcription in cells by blocking the phosphorylation and degradation of I(kappa)B(alpha). purity: 99%
Catalog No. 50-257-6953 Supplier TARGETMOL CHEMICALS INC Supplier No. T338410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Wedelolactone (IKK Inhibitor II) inhibits NF-(kappa)B-mediated gene transcription in cells by blocking the phosphorylation and degradation of I(kappa)B(alpha). purity: 99%