Learn More
Enzo Life Sciences Wedelolactone (1mg). CAS: 524-12-9
SDP
Supplier:  Enzo Life Sciences BMLEI3160001
Encompass_Preferred
Wedelolactone is a naturally occuring furanocoumarin isolated from Eclipta prostrata, an herbal medicine used to treat septic shock in traditional Chinese medicine. It inhibits the IκB kinase (IKK) complex and supresses LPS-induced caspase-11 expression in LPS-treated BALB 3T3 cells (IC50=35µM) Alternative name: 7-Methoxy-5,11,12-trihydroxycoumestan. Purity: ≥96% (HPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0704