Learn More
AdipoGen Wedelolactone (5 mg)
SDP
Supplier:  AdipoGen AGCN20424M005
Encompass_Preferred
Chemical. CAS: 524-12-9. Formula: C16H10O7. MW: 314.2. Isolated from Eclipta prostrata. Cell permeable, selective and irreversible IKKalpha and beta kinase activity inhibitor. NF-kappaB inhibitor. Blocks phosphorylation and degradation of IkappaBalpha. Antihepatotoxic. 5-Lipoxygenase (5-LOX) inhibitor. Anti-inflammatory. HIV-1 integrase inhibitor. Anticancer and antiproliferative compound. Modulates androgen receptor (AR) activation. Apoptosis inducer via downregulation of PKCepsilon. DNA topoisomerase IIalpha inhibitor. G protein-coupled receptor-35 (GPR35) agonist. Inhibits adipogenic differentiation of mesenchymal stem cells (hAMSCs) through ERK pathway. STAT1 protein dephosphorylation inhibitor. Antifibrotic.
Catalog No. 50-205-3594