Learn More
Electron Microscopy Sciences Tripod Polisher Wedge Polishing Mount
SDP
Catalog No. 50980877
Encompass
Includes a Wedge Polishing Clamp (#50116-62) and 5 of Pyrex Wedge Polishing Rod (#50116-63), Holder is used for TEM Wedge Polishing and provides small sample mounting area allowing for maximum adjustment range of the sample.
Catalog No. 50-980-877 Supplier Electron Microscopy Sciences Supplier No. 5011661
Includes a Wedge Polishing Clamp (#50116-62) and 5 of Pyrex Wedge Polishing Rod (#50116-63), Holder is used for TEM Wedge Polishing and provides small sample mounting area allowing for maximum adjustment range of the sample.