Learn More
NEST SCIENTIFIC INC WEDGED IN FUNNELS 100CS
Catalog No. NC2949881
Encompass_Preferred
These funnels feature a tapered wedge shape that allows them to snugly fit into tubes for a leakproof seal. Made from USP grade VI polypropylene. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility.
Catalog No. NC2949881 Supplier NEST SCIENTIFIC INC Supplier No. 203901
These funnels feature a tapered wedge shape that allows them to snugly fit into tubes for a leakproof seal. Made from USP grade VI polypropylene. Molded in ISO 7 cleanroom and lot tested free of DNase, RNAase, DNA, cytoxins/endotoxins and BSE/TSE. Produced in a cGMP (21 CFR part 820/821), ISO 13485 and ISO 9001 facility.