Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Novex™ Tris-Glycine Mini Protein Gels, 10–20%, 1.0 mm, WedgeWell™ format

Catalog No. xp10202box
Encompass_Preferred
Change view
Click to view available options
Wells:
10-well
12-well
15-well
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wells
XP10202BOX 12-well
XP10200BOX 10-well
XP10205BOX 15-well
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. XP10202BOX Supplier Invitrogen™ Supplier No. XP10202BOX
Only null left
Add to Cart
Add to Cart

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity.

Invitrogen Novex Tris-Glycine Mini Gels, WedgeWell format, are polyacrylamide gels that provide reproducible separation of a wide range of proteins into well-resolved bands with two times the loading capacity. Novex Tris-Glycine Mini Gels are based on traditional Laemmli protein electrophoresis that allow the use of Laemmli sample and running buffers.

Features of Novex Tris-Glycine gels, WedgeWell format, include:
Improved shelf life—store gels for up to 21 weeks at 4°C
Diversity—use for native and denaturing protein assays
Wedge-shaped wells—easily load up to two times more sample volume
Fast run conditions—quickly separate your proteins using constant voltage in less than 60 minutes

Choose the right gel format for your experiments
Novex Tris-Glycine gels, WedgeWell format, come in a variety of fixed concentrations from 6% to 18%, as well as gradients with ranges of 4–12%, 4–20%, 8–16%, and 10–20%. You can select from our many well formats, including 10-, 12-, and 15-well, and different pack sizes.

Run your proteins in native or denatured form
Novex Tris-Glycine gels, WedgeWell format, do not contain SDS and can be used to run your proteins in native or in denatured form. For denatured proteins, we recommend using Novex Tris-Glycine SDS Sample Buffer (LC2676) and Novex Tris-Glycine SDS Running Buffer (LC26755). For native proteins, we recommend using Novex Tris-Glycine Native Sample Buffer (LC2673) and Novex Tris-Glycine Native Running Buffer (LC2672).

Convenient to use
Novex Tris-Glycine gels, WedgeWell format, have innovative wedge-shaped wells that allow up to two times more sample load volume than other 1.0 mm-thick gels. The wedge-shaped wells also have larger openings, making sample loading easier.

For transferring your proteins to a membrane, we recommend using Novex Tris-Glycine Transfer Buffer (LC3675) when preforming traditional wet transfer using the Invitrogen Mini Blot Module (B1000). Transfers can also be performed with rapid semi-dry transfer using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 10 to 20%
Gel Size Mini
Gel Thickness 1.0 mm
Gel Type Tris-Glycine Plus
Recommended Applications Denaturing, Native
Sample Loading Volume Up to 45 μL
Separation Range 6 to 200 kDa
Separation Type Denaturing, Native
Wells 12-well
Mode of Separation Molecular Weight
Quantity 10 gels (1 box)
Shipping Condition Wet Ice
Storage Requirements Store at 2°C to 8°C. Do not freeze.
Length (Metric) 8 cm
Width (Metric) 8 cm
Product Line Novex, WedgeWell
For Use With (Equipment) Mini Gel Tank
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.