Learn More
Key Scientific Products WEE-TAB ESC/PYR, 28/PK
SDP
Catalog No. NC0515144
Encompass_Preferred
The ability to hydrolyse both esculin and PYR are valuable tests to identify an assortment of different organisms, in particular Streptococci and Enterococci. Key Scientific has combined these two tests into one easy to use, inexpensive tablet. Wee-Tabs are packaged in 28 ready to use tubes, and have room temperature storage and a two year expiration.
Catalog No. NC0515144 Supplier Key Scientific Products Supplier No. K1137
The ability to hydrolyse both esculin and PYR are valuable tests to identify an assortment of different organisms, in particular Streptococci and Enterococci. Key Scientific has combined these two tests into one easy to use, inexpensive tablet. Wee-Tabs are packaged in 28 ready to use tubes, and have room temperature storage and a two year expiration.