Learn More
Key Scientific Products WEE TAB TREHALOSE
SDP
Catalog No. NC0477398
Encompass_Preferred
WEE-TAB Trehalose is one of the hard to find rare sugars for fermentation tests which normally have a very short shelf life. They are a cost effective way to keep such tests in stock with minimal waste. The small tubes are packaged 28 per container and take up only 3 inches of shelf space. They are useful for many types of organisms and results are able to be seen in 2-4 hours. They are a cost-effective rapid method to referee MALDI or other ID system for equivocal results.
Catalog No. NC0477398 Supplier Key Scientific Products Supplier No. K1640
WEE-TAB Trehalose is one of the hard to find rare sugars for fermentation tests which normally have a very short shelf life. They are a cost effective way to keep such tests in stock with minimal waste. The small tubes are packaged 28 per container and take up only 3 inches of shelf space. They are useful for many types of organisms and results are able to be seen in 2-4 hours. They are a cost-effective rapid method to referee MALDI or other ID system for equivocal results.