Learn More
Key Scientific Products WEE-TAB UREA / IND, 28/PK
SDP
Catalog No. NC0515141
Encompass_Preferred
Traditional urease and indole tests require long incubation and require special handling for anaerobes.� The Urea/Indole Wee-Tab gives results for both Urease and Indole production, incubates aerobically for 2-4 hours, and has 28 ready to use tubes per package.� The tablets are stored at room temperature with a long shelf life. KEY Urea/Indole tests may be used for enterics and other types of bacteria, including anaerobes and Campylobacter species.
Catalog No. NC0515141 Supplier Key Scientific Products Supplier No. K1651
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Traditional urease and indole tests require long incubation and require special handling for anaerobes.� The Urea/Indole Wee-Tab gives results for both Urease and Indole production, incubates aerobically for 2-4 hours, and has 28 ready to use tubes per package.� The tablets are stored at room temperature with a long shelf life. KEY Urea/Indole tests may be used for enterics and other types of bacteria, including anaerobes and Campylobacter species.