Learn More
Carnabio Usa Inc WEE1/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0517710
Encompass
WEE1, Catalytic domain, Wild type, Amino Acid 215-646, NP_003381.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1025