Learn More
Eurofins Discoverx Wee1, active, 250 µg. (14-925M)
Supplier:  Eurofins Discoverx 14925M
Encompass
N-terminal GST-tagged and C-terminal 6xHis-tagged recombinant, human Wee1 amino acids 214-end. Acts as a negative regulator of entry into mitosis (G2 to M transition) by protecting the nucleus from cytoplasmically activated cyclin B1-complexed CDK1 before the onset of mitosis by mediating phosphorylation of CDK1 on 'Tyr-15'. Specifically phosphorylates and inactivates cyclin B1-complexed CDK1 reaching a maximum during G2 phase and a minimum as cells enter M phase. Phosphorylation of cyclin B1-CDK1 occurs exclusively on 'Tyr-15' and phosphorylation of monomeric CDK1 does not occur. Its activity increases during S and G2 phases and decreases at M phase when it is hyperphosphorylated. A correlated decrease in protein level occurs at M/G1 phase, probably due to its degradation. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9424