Learn More
Medchemexpress LLC WEE1-IN-3 | 2272976-28-8 | 98.6% | 497.59 | 100 MG
SDP
Catalog No. 5000349585
Encompass_Preferred
WEE1-IN-3 is a potent inhibitor of Wee1 kinase with an IC50 of less than 10 nM. It demonstrates anticancer activities. WEE1 kinase is crucial for the G2-M cell-cycle checkpoint, which is involved in DNA repair before mitotic entry. While normal cells repair damaged DNA during G1 arrest, cancer cells often have a deficient G1-S checkpoint and rely on a functional G2-M checkpoint for DNA repair. WEE1 is found to be overexpressed in various cancer types. WEE1-IN-3 inhibits cancer cell growth with IC50 values of 100-1000 nM for SW480 cells and less than 100 nM for H23 cells.
  • Potent Wee1 kinase inhibitor with an IC50 of <10 nM.
  • Exhibits anticancer activities.
  • Inhibits cancer cell growth in SW480 and H23 cells.
  • Available as solid or solution in DMSO.
Catalog No. 50-003-49585 Supplier Medchemexpress LLC Supplier No. HY138239100MG
WEE1-IN-3 is a potent inhibitor of Wee1 kinase with an IC50 of less than 10 nM. It demonstrates anticancer activities. WEE1 kinase is crucial for the G2-M cell-cycle checkpoint, which is involved in DNA repair before mitotic entry. While normal cells repair damaged DNA during G1 arrest, cancer cells often have a deficient G1-S checkpoint and rely on a functional G2-M checkpoint for DNA repair. WEE1 is found to be overexpressed in various cancer types. WEE1-IN-3 inhibits cancer cell growth with IC50 values of 100-1000 nM for SW480 cells and less than 100 nM for H23 cells.
  • Potent Wee1 kinase inhibitor with an IC50 of <10 nM.
  • Exhibits anticancer activities.
  • Inhibits cancer cell growth in SW480 and H23 cells.
  • Available as solid or solution in DMSO.