Learn More
Cayman Chemical Wee1 InhIbItr 1mg
SDP
Catalog No. 502913384
Encompass_Preferred
An inhibitor of Wee1 (IC50 = 11 nM); selective for Wee1 over Chk1 (IC50 = 440 nM); inhibits the ATPase activity of LptB (IC50 = 25 µM for the E. coli enzyme)
Catalog No. 50-291-3384 Supplier Cayman Chemical Supplier No. 353911 MG
Add to Cart
Add to Cart
An inhibitor of Wee1 (IC50 = 11 nM); selective for Wee1 over Chk1 (IC50 = 440 nM); inhibits the ATPase activity of LptB (IC50 = 25 µM for the E. coli enzyme)