Learn More
Graphic Controls LLC WEEKLY CIRCULAR CHART 100/BX
Catalog No. NC9558001
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Ink writing circular chart, 100 per each: Chart Size: 09.938" Time Rotation: 7D Number of Ranges: 2 Ranges: (-30) - (70), Non-Uniform -30 TO 0 TO 70 BY 10 (0) - (100), Non-Uniform 0 TO 100 BY 10
Catalog No. NC9558001 Supplier Graphic Controls LLC Supplier No. 31485751
This item has a minimum qty of 2 per supplier requirements.
Ink writing circular chart, 100 per each: Chart Size: 09.938" Time Rotation: 7D Number of Ranges: 2 Ranges: (-30) - (70), Non-Uniform -30 TO 0 TO 70 BY 10 (0) - (100), Non-Uniform 0 TO 100 BY 10