Learn More
Medchemexpress LLC WEHI-345 | 1354825-58-3 | 99.31% | 401.46 | 1 ML
SDP
Catalog No. 5000385267
Encompass_Preferred
WEHI-345 is a potent and selective RIPK2 kinase inhibitor with an IC50 of 0.13 μM. It delays RIPK2 ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.
  • Potent and selective RIPK2 kinase inhibitor.
  • IC50 of 0.13 μM against RIPK2.
  • Delays ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.
Catalog No. 50-003-85267 Supplier Medchemexpress LLC Supplier No. HY1893710MM/1ML
WEHI-345 is a potent and selective RIPK2 kinase inhibitor with an IC50 of 0.13 μM. It delays RIPK2 ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.
  • Potent and selective RIPK2 kinase inhibitor.
  • IC50 of 0.13 μM against RIPK2.
  • Delays ubiquitylation and NF-κB activation on oligomerization domain (NOD) stimulation.