Learn More
Selleck Chemical LLC WEHI-539
SDP
Catalog No. 5000002937
Encompass_Preferred
WEHI-539 has high affinity (IC50 1 1 nM) and selectivity for BCL-XL and potently kills cells by selectively antagonizing its prosurvival activity It has more than a 400-fold higher affinity for BCL-XL versus other prosurvival BCL-2 family members
Catalog No. 50-000-02937 Supplier Selleck Chemical LLC Supplier No. S71001MG
WEHI-539 has high affinity (IC50 1 1 nM) and selectivity for BCL-XL and potently kills cells by selectively antagonizing its prosurvival activity It has more than a 400-fold higher affinity for BCL-XL versus other prosurvival BCL-2 family members