Learn More
Electron Microscopy Sciences Wehnelt Aperture 500 µm Hole
SDP
Catalog No. 50980824
Encompass_Preferred
Made from 0.004" tantalum foil. Replaceable tantalum apertures for some microscopes (Amray and Philips). They improve the performance of the electron gun.
Catalog No. 50-980-824 Supplier Electron Microscopy Sciences Supplier No. 49050TA
Made from 0.004" tantalum foil. Replaceable tantalum apertures for some microscopes (Amray and Philips). They improve the performance of the electron gun.