Learn More
Ethos Biosciences Weigert Iron Hematoxylin Set
Catalog No. 5000819182
Encompass_Preferred
Weigert s Iron Hematoxylin kit is a two-component staining set used to prepare a stable iron-mordanted hematoxylin solution that produces crisp deep blue-black nuclear staining with excellent resistance to decolorization It is a classic progressive nuclear stain widely regarded for its durability in complex staining procedures Mix equal parts of solution A and b immediatley before use
Catalog No. 50-008-19182 Supplier Ethos Biosciences Supplier No. 6260
Weigert s Iron Hematoxylin kit is a two-component staining set used to prepare a stable iron-mordanted hematoxylin solution that produces crisp deep blue-black nuclear staining with excellent resistance to decolorization It is a classic progressive nuclear stain widely regarded for its durability in complex staining procedures Mix equal parts of solution A and b immediatley before use