Learn More
Electron Microscopy Sciences Weigert's Iodide Solution 500 ML
SDP
Catalog No. 5029710
Encompass_Preferred
Weigert's Iodine Solution is a histological stain for the demonstration of connective tissue cases of emphysema, arteriosclerosis and other vascular diseases. It is a ready-to-use, high quality staining solutions.
Catalog No. 50-297-10 Supplier Electron Microscopy Sciences Supplier No. 2604505
Weigert's Iodine Solution is a histological stain for the demonstration of connective tissue cases of emphysema, arteriosclerosis and other vascular diseases. It is a ready-to-use, high quality staining solutions.