Learn More
Electron Microscopy Sciences Weigert's Iodide Solution 1 LT
SDP
Catalog No. 5029711
Encompass_Preferred
Weigert's Iodine Solution is a histological stain for the demonstration of connective tissue cases of emphysema, arteriosclerosis and other vascular diseases. It is a ready-to-use, high quality staining solutions.
Catalog No. 50-297-11 Supplier Electron Microscopy Sciences Supplier No. 2604506
Weigert's Iodine Solution is a histological stain for the demonstration of connective tissue cases of emphysema, arteriosclerosis and other vascular diseases. It is a ready-to-use, high quality staining solutions.