Learn More
Electron Microscopy Sciences Weigert's Iron Hematoxylin A 1 LT
SDP
Catalog No. 5029713
Encompass_Preferred
Weigert's Iron Hematoxylin Stain is used selectivity for nuclei, fungal, and general applications. It is a ready-to-use, high quality staining solution that is packaged and prepared in convenient and precise quantities.
Catalog No. 50-297-13 Supplier Electron Microscopy Sciences Supplier No. 2604406
Weigert's Iron Hematoxylin Stain is used selectivity for nuclei, fungal, and general applications. It is a ready-to-use, high quality staining solution that is packaged and prepared in convenient and precise quantities.