Learn More
Electron Microscopy Sciences Weigert's Iron Hematoxylin B 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2637004
Encompass_Preferred
Used in Weigert's Resorcin Fuchsin for staining elastic fibers in blood vessel walls, nuclei & collagen. See references: Mallory, FB: Pathological Technique; New York: Hafner Publishing Co., c. 1961, p. 168, Clark G: Staining Procedures, Baltimore: Williams & Wilkins Co., 3rd ed. c. 1973, p.60 and Luna, I. G.: AFIP Histologic Staining Methods: New York: McGraw Hill Book Co., 3rd ed., c. 1968, p. 80
Catalog No. 50-317-84