Learn More
Electron Microscopy Sciences Weigert's Iron Hematoxylin B 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2604415
Encompass_Preferred
Weigert's Iron Hematoxylin Stain is used selectivity for nuclei, fungal, and general applications. It is a ready-to-use, high quality staining solution that is packaged and prepared in convenient and precise quantities.
Catalog No. 50-297-15