Learn More
MilliporeSigma™ Weigert's Iron Hematoxylin kit
Catalog No. M1159730002 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 x 500 mL
Catalog No. Quantity
M1159730002 2 x 500 mL
1 options
Catalog No. M1159730002 Supplier MilliporeSigma™ Supplier No. 1.15973.0002
Add to Cart
Add to Cart
For staining nuclei of tissue sections
  • Elastic fibers stained according to van Gieson method
  • Connective tissue stained by various trichromo staining techniques, where acidic stains were used as counter-stains
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stain Kit
Quantity 2 x 500 mL
Solution Type Stain Kit
Form Liquid