Learn More
Polysciences, Inc. WEIGERTS HEMATOXYLIN KIT (A&B)
SDP
Catalog No. NC0856208
Encompass_Preferred
A nuclear staining solution containing, hematoxylin, ferric chloride and hydrochloric acid used in many non-routine techniques (special stains) because, it resists decolorization in acidic staining solutions. Ferric Chloride is a strong oxidizer, so it serves both as a mordant and oxidizer for Weigert's Hematoxylin.250ml
Catalog No. NC0856208 Supplier Polysciences, Inc. Supplier No. 253731
A nuclear staining solution containing, hematoxylin, ferric chloride and hydrochloric acid used in many non-routine techniques (special stains) because, it resists decolorization in acidic staining solutions. Ferric Chloride is a strong oxidizer, so it serves both as a mordant and oxidizer for Weigert's Hematoxylin.250ml