Learn More
Electron Microscopy Sciences Weigerts Iron Hematoxylin Solution A 500 ML
SDP
Catalog No. 5031666
Encompass_Preferred
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.
Catalog No. 50-316-66 Supplier Electron Microscopy Sciences Supplier No. 261091A
Used in Gridley's Method for Endamoeba Histolytica. For bacteria stain: Amoebae, their nuclei and ingested erythrocytes.